Learn More
Ethyl 5-bromonicotinate, 98%
Description
Ethyl 5-bromonicotinate is used as a key starting material for the preparation of 5-bromonicotinic acid hydrazide via reaction with a hydrazine hydrate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ethyl 5-bromonicotinate is used as a key starting material for the preparation of 5-bromonicotinic acid hydrazide via reaction with a hydrazine hydrate.
Notes
Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 39°C to 42°C |
| Boiling Point | 86°C to 92°C (0.5 mmHg) |
| Quantity | 5 g |
| Beilstein | 131473 |
| Formula Weight | 230.07 |
| Percent Purity | 98% |
| Chemical Name or Material | Ethyl 5-bromonicotinate |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.