missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-bromothiophene-2-carboxylate, 99%

Product Code. 11486076
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11486076 1 g 1g
11496076 5 g 5g
11406086 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11486076 Supplier Thermo Scientific Alfa Aesar Supplier No. B22355.03
 Restricted Item
Estimated Shipment: 20-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 5751-83-7 | C7H7BrO2S | 235.095 g/mol

Ethyl 5-bromothiophene-2-carboxylate is used as an intermediate in organic syntheses

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 5-bromothiophene-2-carboxylate is used as an intermediate in organic syntheses

Solubility
Slightly soluble in water

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5751-83-7
Molecular Formula C7H7BrO2S
Molecular Weight (g/mol) 235.095
MDL Number MFCD02683089
InChI Key PZNHMXAOMDQLLE-UHFFFAOYSA-N
Synonym 5-bromothiophene-2-carboxylic acid ethyl ester, ethyl 5-bromo-2-thiophenecarboxylate, ethyl5-bromothiophene-2-carboxylate, 5-bromo-thiophene-2-carboxylic acid ethyl ester, 2-thiophenecarboxylic acid, 5-bromo-, ethyl ester, pubchem16118, acmc-209zz5, ksc494s9b, 2-bromo-5-ethoxycarbonylthiophene, ethyl 5-bromothiophene-2carboxylate
PubChem CID 605723
IUPAC Name ethyl 5-bromothiophene-2-carboxylate
SMILES CCOC(=O)C1=CC=C(S1)Br

Specifications

Boiling Point 94°C to 96°C (4 mmHg)
Quantity 1 g
Solubility Information Slightly soluble in water
Formula Weight 235.1
Percent Purity 99%
Chemical Name or Material Ethyl 5-bromothiophene-2-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.