missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 6-amino-5-methoxyindole-2-carboxylate, 95%

Product Code. 11407236
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11407236 1 g 1g
11417236 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11407236 Supplier Thermo Scientific Alfa Aesar Supplier No. B25549.03
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 107575-60-0 | C12H14N2O3 | 234.255 g/mol

Ethyl 6-amino-5-methoxyindole-2-carboxylate is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 6-amino-5-methoxyindole-2-carboxylate is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in dichloromethane, ethyl acetate and methanol.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107575-60-0
Molecular Formula C12H14N2O3
Molecular Weight (g/mol) 234.255
MDL Number MFCD08064224
InChI Key DIEUGINJYISDCB-UHFFFAOYSA-N
PubChem CID 10966428
IUPAC Name ethyl 6-amino-5-methoxy-1H-indole-2-carboxylate
SMILES CCOC(=O)C1=CC2=CC(=C(C=C2N1)N)OC

Specifications

Melting Point 167°C to 169°C
Quantity 1 g
Synonym ethyl 6-amino-5-methoxyindole-2-carboxylate, acmc-209zno, ethyl6-amino-5-methoxy-1h-indole-2-carboxylate, ethyl 6-amino-5-methoxy-1h-indole-2-carboxylate, 1h-indole-2-carboxylicacid, 6-amino-5-methoxy-, ethyl ester
Solubility Information Soluble in dichloromethane,ethyl acetate and methanol.
Formula Weight 234.25
Percent Purity 95%
Chemical Name or Material Ethyl 6-amino-5-methoxyindole-2-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.