missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 6-amino-5-methoxyindole-2-carboxylate, 95%

Product Code. p-7028309
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11407236

Brand: Thermo Scientific Alfa Aesar B25549.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 107575-60-0 | C12H14N2O3 | 234.255 g/mol

Ethyl 6-amino-5-methoxyindole-2-carboxylate is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 6-amino-5-methoxyindole-2-carboxylate is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in dichloromethane, ethyl acetate and methanol.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107575-60-0
Molecular Formula C12H14N2O3
Molecular Weight (g/mol) 234.255
MDL Number MFCD08064224
InChI Key DIEUGINJYISDCB-UHFFFAOYSA-N
PubChem CID 10966428
IUPAC Name ethyl 6-amino-5-methoxy-1H-indole-2-carboxylate
SMILES CCOC(=O)C1=CC2=CC(=C(C=C2N1)N)OC

Specifications

Melting Point 167°C to 169°C
Quantity 1 g
Synonym ethyl 6-amino-5-methoxyindole-2-carboxylate, acmc-209zno, ethyl6-amino-5-methoxy-1h-indole-2-carboxylate, ethyl 6-amino-5-methoxy-1h-indole-2-carboxylate, 1h-indole-2-carboxylicacid, 6-amino-5-methoxy-, ethyl ester
Solubility Information Soluble in dichloromethane,ethyl acetate and methanol.
Formula Weight 234.25
Percent Purity 95%
Chemical Name or Material Ethyl 6-amino-5-methoxyindole-2-carboxylate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.