missing translation for 'onlineSavingsMsg'
Learn More

Ethyl butyrylacetate, 98%

Product Code. 11445133
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
This item is not returnable. View return policy

Product Code. 11445133

Brand: Thermo Scientific Alfa Aesar A12786.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3249-68-1 | C8H14O3 | 158.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3249-68-1
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.20
MDL Number MFCD00009401
InChI Key KQWWVLVLVYYYDT-UHFFFAOYSA-N
Synonym ethyl butyrylacetate, ethyl butyroacetate, hexanoic acid, 3-oxo-, ethyl ester, ethyl beta-ketohexanoate, ethyl alpha-butyrylacetate, 3-ketohexanoic acid ethyl ester, ethyl-3-oxohexanoate, unii-8q1ahg710e, ethyl .alpha.-butyrylacetate, fema no. 3683
PubChem CID 238498
ChEBI CHEBI:18119
IUPAC Name ethyl 3-oxohexanoate
SMILES CCCC(=O)CC(=O)OCC

Specifications

Melting Point -44°C
Density 0.99
Boiling Point 94°C to 95°C (12 mmHg)
Flash Point 78°C (172°F)
Refractive Index 1.427
Quantity 50 g
Beilstein 507689
Formula Weight 158.2
Percent Purity 98%
Chemical Name or Material Ethyl butyrylacetate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.