missing translation for 'onlineSavingsMsg'
Learn More

Ethyl coumarin-3-carboxylate, 98%

Product Code. 11414254
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11414254 5 g 5g
11404254 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11414254 Supplier Thermo Scientific Alfa Aesar Supplier No. A12553.06
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1846-76-0 | C12H10O4 | 218.208 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 1846-76-0
Molecular Formula C12H10O4
Molecular Weight (g/mol) 218.208
MDL Number MFCD00016964
InChI Key XKHPEMKBJGUYCM-UHFFFAOYSA-N
Synonym ethyl coumarin-3-carboxylate, ethyl 2-oxo-2h-chromene-3-carboxylate, 3-ethoxycarbonylcoumarin, ethyl 3-coumarincarboxylate, 3-carbethoxycoumarin, ethylcoumarin-3-carboxylate, coumarin-3-carboxylic acid ethyl ester, 1,2-benzopyran-2-one-3-carboxylic acid, ethyl ester, 2h-1-benzopyran-3-carboxylic acid, 2-oxo-, ethyl ester, 2h-1-benzopyran-3-carboxylicacid, 2-oxo-, ethyl ester
PubChem CID 15800
IUPAC Name ethyl 2-oxochromene-3-carboxylate
SMILES CCOC(=O)C1=CC2=CC=CC=C2OC1=O

Spezifikation

Melting Point 92°C to 95°C
Quantity 5 g
Beilstein 200147
Formula Weight 218.21
Percent Purity 98%
Chemical Name or Material Ethyl coumarin-3-carboxylate

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.