Learn More
Ethyl diethoxyacetate, 97+%
Description
Ethyl diethoxyacetate is used in the preparation of a fatty acid analog containing DTS, 12,13-dioxotetradecanoic acid di(N-methyl-thiosemicarbazone) (FA-DTS). It is used for synthesis of heterocyclic compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ethyl diethoxyacetate is used in the preparation of a fatty acid analog containing DTS, 12,13-dioxotetradecanoic acid di(N-methyl-thiosemicarbazone) (FA-DTS). It is used for synthesis of heterocyclic compounds.
Solubility
Soluble in Dichloromethane, Ethyl Acetate and Methanol.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications
Specifications
| Density | 0.988 |
| Boiling Point | 198°C to 200°C |
| Flash Point | 85°C (185°F) |
| Refractive Index | 1.41 |
| Quantity | 25 g |
| Beilstein | 878579 |
| Solubility Information | Soluble in Dichloromethane,Ethyl Acetate and Methanol. |
| Formula Weight | 176.21 |
| Percent Purity | ≥97% |
| Chemical Name or Material | Ethyl diethoxyacetate |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.