missing translation for 'onlineSavingsMsg'
Learn More

Ethyl hippurate, 96%

Product Code. 11456486
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11456486 5 g 5g
11466486 25 g 25g
11476486 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11456486 Supplier Thermo Scientific Alfa Aesar Supplier No. B24765.06
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 1499-53-2 | C11H13NO3 | 207.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1499-53-2
Molecular Formula C11H13NO3
Molecular Weight (g/mol) 207.23
MDL Number MFCD00026890
InChI Key PTXRQIPIELXJFH-UHFFFAOYSA-N
Synonym ethyl hippurate, ethyl n-benzoylglycinate, n-benzoylglycine ethyl ester, ethyl 2-phenylformamido acetate, benzoic amide, n-ethoxycarbonyl methyl, ethylhippurate, ethyl benzoylaminoacetate, acmc-1buir, hippuric acid ethyl ester, maybridge1_006087
PubChem CID 226558
IUPAC Name ethyl 2-benzamidoacetate
SMILES CCOC(=O)CNC(=O)C1=CC=CC=C1

Specifications

Melting Point 58°C to 60°C
Quantity 5 g
Formula Weight 207.23
Percent Purity 98%
Chemical Name or Material Ethyl hippurate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.