missing translation for 'onlineSavingsMsg'
Learn More

Ethyl iodoacetate, 98%, Thermo Scientific Chemicals

Product Code. 11471207
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11471207 100 g 100g
11451207 5 g 5g
11461207 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11471207 Supplier Thermo Scientific Alfa Aesar Supplier No. B24177.22
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
This item is not returnable. View return policy

Ethyl iodoacetate was used in the synthesis of trans-2,3-disubstituted indolines by reacting with 1-azido-2-allylbenzene derivatives via a diastereoselective radical cascade.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl iodoacetate was used in the synthesis of trans-2,3-disubstituted indolines by reacting with 1-azido-2-allylbenzene derivatives via a diastereoselective radical cascade.

Solubility
Insoluble in water.

Notes
Light sensitive. Store in dark. Storage temperature is 2-8°C. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-48-3
Molecular Formula C4H7IO2
Molecular Weight (g/mol) 214.00
MDL Number MFCD00001081
InChI Key MFFXVVHUKRKXCI-UHFFFAOYSA-N
Synonym ethyl iodoacetate, acetic acid, iodo-, ethyl ester, ethyl monoiodoacetate, iodoacetic acid, ethyl ester, acetic acid, 2-iodo-, ethyl ester, ethylester kyseliny jodoctove czech, acetic acid, 2-iodo-,ethyl ester, ethylester kyseliny jodoctove, ethyliodoacetate, iodoacetic acid ethyl
PubChem CID 12183
IUPAC Name ethyl 2-iodoacetate
SMILES CCOC(=O)CI

Specifications

Boiling Point 179°C to 180°C
Flash Point 76°C (169°F)
Quantity 100 g
UN Number UN2927
Beilstein 741934
Formula Weight 214
Percent Purity 98%
Chemical Name or Material Ethyl iodoacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.