missing translation for 'onlineSavingsMsg'
Learn More

Ethyl maltol, 98%

Product Code. 15254258 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15254258 25 g 25g
15264258 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15254258 Supplier Thermo Scientific Acros Supplier No. 450800250
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4940-11-8 | C7H8O3 | 140.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11-8-4940
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.14

Specifications

Melting Point 89.0°C to 93.0°C
Boiling Point 289.0°C to 290.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 25 g
Merck Index 15,3879
Synonym ethyl maltol, 2-ethyl-3-hydroxy-4h-pyran-4-one, 2-ethyl-3-hydroxy-4-pyrone, veltol plus, 3-hydroxy-2-ethyl-4-pyrone, ethylmaltol, 2-ethylpyromeconic acid, 4h-pyran-4-one, 2-ethyl-3-hydroxy, 3-hydroxy-2-ethyl-1,4-pyrone, 2-ethyl pyromeconic acid
InChI Key YIKYNHJUKRTCJL-UHFFFAOYSA-N
SMILES CCC1=C(C(=O)C=CO1)O
IUPAC Name 2-ethyl-3-hydroxypyran-4-one
PubChem CID 21059
Formula Weight 140.14
Percent Purity 98%
Chemical Name or Material Ethyl maltol
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl maltol, 98%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.