missing translation for 'onlineSavingsMsg'
Learn More

Ethyl mesitylglyoxylate, 97%

Product Code. 11327896
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11327896 25 g 25g
11317896 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11327896 Supplier Thermo Scientific Alfa Aesar Supplier No. L13106.14
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 5524-57-2 | C13H16O3 | 220.268 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5524-57-2
Molecular Formula C13H16O3
Molecular Weight (g/mol) 220.268
MDL Number MFCD00151821
InChI Key XRSPPFOBPRPYLD-UHFFFAOYSA-N
Synonym ethyl mesitylglyoxylate, ethyl 2-mesityl-2-oxoacetate, ethyl 2-oxo-2-2,4,6-trimethylphenyl acetate, acmc-1asq7, mesitylglyoxylic acid ethyl ester, ethyl 2,4,6-trimethylphenyl glyoxylate, alpha-oxo-2,4,6-trimethylbenzeneacetic acid ethyl ester
PubChem CID 2736481
IUPAC Name ethyl 2-oxo-2-(2,4,6-trimethylphenyl)acetate
SMILES CCOC(=O)C(=O)C1=C(C=C(C=C1C)C)C

Specifications

Boiling Point 164°C to 165°C (10 mmHg)
Refractive Index 1.512
Quantity 25 g
Beilstein 2970031
Sensitivity Moisture sensitive
Formula Weight 220.27
Percent Purity 97%
Chemical Name or Material Ethyl mesitylglyoxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.