missing translation for 'onlineSavingsMsg'
Learn More

Ethyl methyl sulfide, 97%

Product Code. 11377875
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11377875 25 g 25g
11387875 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11377875 Supplier Thermo Scientific Alfa Aesar Supplier No. L02859.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 624-89-5 | C3H8S | 76.16 g/mol

Ethyl Methyl Sulfide is used as a corrosion inhibitor for zinc metal which finds wide applications like electrode material in batteries, sacrificial anodes and metallic coatings. It is used in mixtures designed to odorize natural gas and is made to by-pass injection system.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl Methyl Sulfide is used as a corrosion inhibitor for zinc metal which finds wide applications like electrode material in batteries, sacrificial anodes and metallic coatings. It is used in mixtures designed to odorize natural gas and is made to by-pass injection system.

Solubility
Miscible with alcohols and oils. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 624-89-5
Molecular Formula C3H8S
Molecular Weight (g/mol) 76.16
MDL Number MFCD00009268
InChI Key WXEHBUMAEPOYKP-UHFFFAOYSA-N
Synonym ethyl methyl sulfide, methylthioethane, ethane, methylthio, methyl ethyl sulfide, ethyl methyl sulphide, methyl ethyl sulphide, sulfide, ethyl methyl, unii-mr1gzs62tm, methylthio ethane, methylsulfanyl ethane
PubChem CID 12230
IUPAC Name methylsulfanylethane
SMILES CCSC

Specifications

Melting Point -106°C
Density 0.841
Boiling Point 66°C to 67°C
Flash Point −15°C (5°F)
Odor Stench
Linear Formula CH3CH2SCH3
Refractive Index 1.4392
Quantity 25 g
UN Number UN1993
Beilstein 1696871
Solubility Information Miscible with alcohols and oils. Immiscible with water.
Formula Weight 76.16
Percent Purity 97%
Chemical Name or Material Ethyl methyl sulfide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.