missing translation for 'onlineSavingsMsg'
Learn More

Ethyl nitroacetate, 98%

Product Code. 11914941 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11914941 100 g 100g
10566821 5 g 5g
10718352 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11914941 Supplier Thermo Scientific Acros Supplier No. 185171000
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 626-35-7 | C4H7NO4 | 133.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-35-7
Molecular Formula C4H7NO4
Molecular Weight (g/mol) 133.1
InChI Key FTKASJMIPSSXBP-UHFFFAOYSA-N
Synonym ethyl nitroacetate, nitroacetic acid ethyl ester, acetic acid, nitro-, ethyl ester, nitro ethyl acetate, ethylnitroacetate, acetic acid, 2-nitro-, ethyl ester, ethylnitroacetat, ethyinitroacetate, ethyl-2-nitroacetate, zlchem 269
PubChem CID 69379
IUPAC Name ethyl 2-nitroacetate
SMILES CCOC(=O)C[N+](=O)[O-]

Specifications

Color Colorless to Yellow
Density 1.1990g/mL
Boiling Point 105.0°C to 107.0°C (25.0 mmHg)
Flash Point 92°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula O2NCH2CO2C2H5
Refractive Index 1.4235 to 1.4255
Quantity 100 g
Beilstein 02,225
Specific Gravity 1.199
Formula Weight 133.1
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl nitroacetate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl nitroacetate
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.