missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Paraben, TRC

Product Code. 30010129
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010129 5 g 5g
30006437 10 g 10g
17948794 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010129 Supplier Toronto Research Chemicals Supplier No. TRCE9254755G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-47-8
Molecular Formula C9 H10 O3
Molecular Weight (g/mol) 166.17
Synonym Ethyl Parahydroxybenzoate, Ethyl 4-Hydroxybenzoate, Butyl Parahydroxybenzoate Imp. C (EP), Propyl Parahydroxybenzoate Imp. C (EP), Methyl Parahydroxybenzoate Imp. B (EP), 4-(Ethoxycarbonyl)phenol, 4-Carbethoxyphenol, 4-Hydroxybenzoic acid ethyl ester, Aseptin A, Aseptine A, Aseptoform E, Bonomold OE, Easeptol, Ethyl 4-hydroxybenzoate, Ethyl Butex, Ethyl nipagin, Ethyl p-hydroxybenzoate, Ethyl parasept, Ethylparaben, Mekkings E, Mycocten, NSC 23514, NSC 8510, Napagin A, Nipagin A, Nipazin A, Sobrol A, Solbrol A, Tegosept E, p-(Ethoxycarbonyl)phenol, p-Carbethoxyphenol, p-Hydroxybenzoate ethyl ester, p-Hydroxybenzoic acid ethyl ester
IUPAC Name ethyl 4-hydroxybenzoate
SMILES CCOC(=O)c1ccc(O)cc1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C9H10O3/c1-2-12-9(11)7-3-5-8(10)6-4-7/h3-6,10H,2H2,1H3
Formula Weight 166.063
Percent Purity >95%
Chemical Name or Material Ethyl Parahydroxybenzoate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.