missing translation for 'onlineSavingsMsg'
Learn More

Ethyl phenoxyacetate, 98%, Thermo Scientific™

Product Code. 10183581 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10183581 5 g 5g
10491373 25 g 25g
15229876 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10183581 Supplier Thermo Scientific Acros Supplier No. 334940050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 2555-49-9
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.2
MDL Number MFCD00026895
InChI Key MGZFVSUXQXCEHM-UHFFFAOYSA-N
Synonym ethyl phenoxyacetate, acetic acid, phenoxy-, ethyl ester, phenoxyacetic acid ethyl ester, acetic acid, 2-phenoxy-, ethyl ester, phenoxy-acetic acid ethyl ester, phenoxyacetic acid, ethyl ester, ethyl2-phenoxyacetate, pubchem12485, acmc-1ckja, ethylphenoxyacetate
PubChem CID 17365
IUPAC Name ethyl 2-phenoxyacetate
SMILES CCOC(=O)COC1=CC=CC=C1

Specifications

Color Yellow
Density 1.1000g/mL
Boiling Point 136°C (19.0 mmHg)
Flash Point >113°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.5035
Quantity 5 g
Specific Gravity 1.1
Solubility Information Solubility in water: immiscible in water
Formula Weight 180.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl phenoxyacetate, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.