missing translation for 'onlineSavingsMsg'
Learn More

Ethyl phenoxyacetate, 99%

Product Code. 11438136
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11438136 10 g 10g
11448136 50 g 50g
11452197 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11438136 Supplier Thermo Scientific Alfa Aesar Supplier No. A14156.09
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 2555-49-9 | C10H12O3 | 180.203 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2555-49-9
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00026895
InChI Key MGZFVSUXQXCEHM-UHFFFAOYSA-N
Synonym ethyl phenoxyacetate, acetic acid, phenoxy-, ethyl ester, phenoxyacetic acid ethyl ester, acetic acid, 2-phenoxy-, ethyl ester, phenoxy-acetic acid ethyl ester, phenoxyacetic acid, ethyl ester, ethyl2-phenoxyacetate, pubchem12485, acmc-1ckja, ethylphenoxyacetate
PubChem CID 17365
IUPAC Name ethyl 2-phenoxyacetate
SMILES CCOC(=O)COC1=CC=CC=C1

Specifications

Density 1.108
Boiling Point 251°C to 253°C
Flash Point >110°C (230°F)
Refractive Index 1.5055
Quantity 10 g
Beilstein 1871712
Formula Weight 180.2
Percent Purity 99%
Chemical Name or Material Ethyl phenoxyacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.