missing translation for 'onlineSavingsMsg'
Learn More

Ethyl phenylglyoxylate, 98%

Product Code. 11480878
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11480878 100 g 100g
11460878 5 g 5g
11470878 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11480878 Supplier Thermo Scientific Alfa Aesar Supplier No. B25292.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1603-79-8 | C10H10O3 | 178.19 g/mol

Ethyl phenylglyoxylate has been used in preparation of 1,5-dihydro-5-deazaflavin derivatives possessing a chiral substituent at N(3) position.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl phenylglyoxylate has been used in preparation of 1,5-dihydro-5-deazaflavin derivatives possessing a chiral substituent at N(3) position.

Solubility
Soluble in water 1143 mg/L @ 25°C .

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1603-79-8
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.19
MDL Number MFCD00009120
InChI Key QKLCQKPAECHXCQ-UHFFFAOYSA-N
Synonym ethyl benzoylformate, ethyl phenylglyoxylate, ethyl oxophenylacetate, ethylbenzoylformate, phenylglyoxylic acid ethyl ester, ethyl oxo phenyl acetate, benzeneacetic acid, .alpha.-oxo-, ethyl ester, benzoylformic acid ethyl ester, ethyl benzoyl formate, glyoxylic acid, phenyl-, ethyl ester
PubChem CID 15349
ChEBI CHEBI:84260
IUPAC Name ethyl 2-oxo-2-phenylacetate
SMILES CCOC(=O)C(=O)C1=CC=CC=C1

Specifications

Density 1.122
Boiling Point 138°C to 139°C (18 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.516
Quantity 100 g
Sensitivity Moisture sensitive
Solubility Information Soluble in water 1143mg/L @ 25°C .
Formula Weight 178.19
Percent Purity 98%
Chemical Name or Material Ethyl phenylglyoxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.