missing translation for 'onlineSavingsMsg'
Learn More

Ethyl salicylate, 99%

Product Code. 11459332
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11459332 500 g 500g
11449332 100 g 100g
11431870 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11459332 Supplier Thermo Scientific Alfa Aesar Supplier No. A10101.36
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 118-61-6 | C9H10O3 | 166.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-61-6
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00002215
InChI Key GYCKQBWUSACYIF-UHFFFAOYSA-N
Synonym ethyl salicylate, sal ethyl, mesotol, salotan, salicylic ether, salicylic acid, ethyl ester, benzoic acid, 2-hydroxy-, ethyl ester, ethyl o-hydroxybenzoate, o-ethoxycarbonyl phenol, 2-hydroxybenzoic acid ethyl ester
PubChem CID 8365
IUPAC Name ethyl 2-hydroxybenzoate
SMILES CCOC(=O)C1=CC=CC=C1O

Specifications

Melting Point 1°C
Density 1.13
Boiling Point 232°C
Flash Point 107°C (224°F)
Refractive Index 1.522
Quantity 500 g
Beilstein 907659
Merck Index 14,3850
Formula Weight 166.18
Percent Purity 99%
Chemical Name or Material Ethyl salicylate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.