missing translation for 'onlineSavingsMsg'
Learn More

Ethyl salicylate, 99%

Product Code. p-7080537
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11449332

Brand: Thermo Scientific Alfa Aesar A10101.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 118-61-6 | C9H10O3 | 166.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-61-6
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00002215
InChI Key GYCKQBWUSACYIF-UHFFFAOYSA-N
Synonym ethyl salicylate, sal ethyl, mesotol, salotan, salicylic ether, salicylic acid, ethyl ester, benzoic acid, 2-hydroxy-, ethyl ester, ethyl o-hydroxybenzoate, o-ethoxycarbonyl phenol, 2-hydroxybenzoic acid ethyl ester
PubChem CID 8365
IUPAC Name ethyl 2-hydroxybenzoate
SMILES CCOC(=O)C1=CC=CC=C1O

Specifications

Melting Point 1°C
Density 1.13
Boiling Point 232°C
Flash Point 107°C (224°F)
Refractive Index 1.522
Quantity 100 g
Beilstein 907659
Merck Index 14,3850
Formula Weight 166.18
Percent Purity 99%
Chemical Name or Material Ethyl salicylate
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.