missing translation for 'onlineSavingsMsg'
Learn More

Ethyl salicylate, 99+%, Thermo Scientific Chemicals

Product Code. 10329393 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10329393 25 g Glass bottle 25g
10097691 5 g Glass bottle 5g
10428680 250 g Glass bottle 250g
10041840 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10329393 Supplier Thermo Scientific Acros Supplier No. 150490250
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-61-6
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00002215
InChI Key GYCKQBWUSACYIF-UHFFFAOYSA-N
Synonym ethyl salicylate, sal ethyl, mesotol, salotan, salicylic ether, salicylic acid, ethyl ester, benzoic acid, 2-hydroxy-, ethyl ester, ethyl o-hydroxybenzoate, o-ethoxycarbonyl phenol, 2-hydroxybenzoic acid ethyl ester
PubChem CID 8365
IUPAC Name ethyl 2-hydroxybenzoate
SMILES CCOC(=O)C1=CC=CC=C1O

Specifications

Acidity 1mg KOH/g max.
Melting Point 2.0°C to 3.0°C
Color Colorless to Yellow
Density 1.1280g/mL
Boiling Point 234.0°C
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula 2-(HO)C6H4CO2C2H5
Refractive Index 1.5210 to 1.5250
Quantity 25 g
Beilstein 10,73
Merck Index 15,3902
Specific Gravity 1.128
Solubility Information Solubility in water: slightly soluble. Other solubilities: miscible with alcohol and ether
Formula Weight 166.18
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Ethyl salicylate, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.