missing translation for 'onlineSavingsMsg'
Learn More

Ethyl vinyl ketone, 97%, stab.

Product Code. 11386996
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11386996 5 g 5g
11396996 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11386996 Supplier Thermo Scientific Alfa Aesar Supplier No. L06829.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1629-58-9 | C5H8O | 84.118 g/mol

Ethyl vinyl ketone is used as reagent for annulation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl vinyl ketone is used as reagent for annulation.

Solubility
Insoluble in water; soluble in most organic solvents.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1629-58-9
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.118
MDL Number MFCD00009316
InChI Key JLIDVCMBCGBIEY-UHFFFAOYSA-N
Synonym 1-penten-3-one, ethyl vinyl ketone, ethylvinylketone, 1-pentene-3-one, ethylvinyl ketone, pentenone, ketone, ethyl vinyl, vinyl ethyl ketone, unii-r0053y1az7, fema no. 3382
PubChem CID 15394
IUPAC Name pent-1-en-3-one
SMILES CCC(=O)C=C

Specifications

Density 0.844
Boiling Point 103°C to 104°C
Flash Point −6°C (21°F)
Odor Pungent
Refractive Index 1.42
Quantity 5 g
UN Number UN3286
Beilstein 1735857
Solubility Information Insoluble in water; soluble in most organic solvents.
Formula Weight 84.12
Percent Purity 97%
Chemical Name or Material Ethyl vinyl ketone, stabilized
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.