missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Violet

Product Code. p-7042844
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 15406109

Brand: Thermo Scientific Alfa Aesar J63951.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2390-59-2 | C31H42ClN3 | 492.15 g/mol

Ethyl violet has been used to stain pancreatic tissue and elastin in the detection of gram negative bacteria, and as a reagent for the extractive-spectrophotometric determination of anionic surfactants. It has been used in the extraction and spectrophotometric determination of copper. Along with the anionic dye zincon, ethyl violet is used in counterion dye of proteins in SDS-polyacrylamide gel electrophoresis. Along with the anionic dye zincon, ethyl violet is used in counterion dye of proteins in SDS-polyacrylamide gel electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl violet has been used to stain pancreatic tissue and elastin in the detection of gram negative bacteria, and as a reagent for the extractive-spectrophotometric determination of anionic surfactants. It has been used in the extraction and spectrophotometric determination of copper. Along with the anionic dye zincon, ethyl violet is used in counterion dye of proteins in SDS-polyacrylamide gel electrophoresis. Along with the anionic dye zincon, ethyl violet is used in counterion dye of proteins in SDS-polyacrylamide gel electrophoresis.

Solubility
Soluble in water (10 mg/ml).

Notes
Dye content approx. 80%
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2390-59-2
Molecular Formula C31H42ClN3
Molecular Weight (g/mol) 492.15
MDL Number MFCD00011765
InChI Key JVICFMRAVNKDOE-UHFFFAOYSA-M
Synonym C.I. 42600; Basic Violet 4
PubChem CID 16955
IUPAC Name [4-[bis[4-(diethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-diethylazanium;chloride
SMILES [Cl-].CCN(CC)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC)C1=CC=C(C=C1)N(CC)CC

Specifications

Beilstein 4117566
Solubility Information Soluble in water (10mg/ml).
Formula Weight 492.15
Color Green
Quantity 25 g
Physical Form Powder
Chemical Name or Material Ethyl Violet

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.