missing translation for 'onlineSavingsMsg'
Learn More

ethyle7-hydroxy-4-oxo-4H-chromene-2-carboxylate, 97%, Thermo Scientific™

Product Code. 10532092
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10532092 10 g Amber Glass Bottle 10g
10354222 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10532092 Supplier Thermo Scientific Maybridge Supplier No. BTB10085EA
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 23866-72-0
Molecular Formula C12H10O5
Molecular Weight (g/mol) 234.21
MDL Number MFCD00100382
InChI Key RKIMFICEWCXBCE-UHFFFAOYSA-N
Synonym ethyl 7-hydroxy-4-oxo-4h-chromene-2-carboxylate, 4h-1-benzopyran-2-carboxylic acid, 7-hydroxy-4-oxo-, ethyl ester, chembl82749, 2-ethoxycarbonyl-7-hydroxychromone, ethyl 7-hydroxy-4-oxo-4h-chromen-2-carboxylate, 7-hydroxy-4-oxo-4h-1-benzopyran-2-carboxylic acid ethyl ester, 7-hydroxy-4-oxo-4h-chromene-2-carboxylic acid ethyl ester, 4h-1-benzopyran-2-carboxylicacid, 7-hydroxy-4-oxo-, ethyl ester, maybridge1_002082, 7-hydroxy-4-oxo-4h-chromen-2-carboxylic acid ethyl ester
PubChem CID 5376802
IUPAC Name ethyl 7-hydroxy-4-oxochromene-2-carboxylate
SMILES CCOC(=O)C1=CC(=O)C2=CC=C(O)C=C2O1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 234.21
Percent Purity 97%
Chemical Name or Material Ethyl 7-hydroxy-4-oxo-4H-chromene-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.