missing translation for 'onlineSavingsMsg'
Learn More

Ethylene diacrylate, 95%, stabilized

Product Code. 10314692 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10314692 25 mL Glass bottle 25mL
10562612 5 mL Glass bottle 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10314692 Supplier Thermo Scientific Supplier No. 366970250
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2274-11-5 | C8H10O4 | 170.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2274-11-5
Molecular Formula C8H10O4
Molecular Weight (g/mol) 170.17
MDL Number MFCD00008629
InChI Key KUDUQBURMYMBIJ-UHFFFAOYSA-N
SMILES C=CC(=O)OCCOC(=O)C=C

Specifications

Color Colorless to Yellow
Density 1.0400g/mL
Boiling Point 62.0°C to 65.0°C (2.0 mmHg)
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4520 to 1.4560
Quantity 25 mL
Beilstein 02, 1469
Specific Gravity 1.04
Synonym ethylene diacrylate, ethylene acrylate, ethylene glycol diacrylate, acrylic acid, ethylene ester, acrylic acid, ethylene glycol diester, 2-propenoic acid, 1,2-ethanediyl ester, acrylic acid, diester with ethylene glycol, 1,2-ethanediyl diacrylate, ethylene glycol dipropenoate, unii-42q902riye
Solubility Information Solubility in water: slightly soluble
IUPAC Name 2-prop-2-enoyloxyethyl prop-2-enoate
Stabilizer 100ppm MEHQ
PubChem CID 75282
Formula Weight 170.17
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Ethylene diacrylate, 90%
Mostrar más Mostrar menos

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.