missing translation for 'onlineSavingsMsg'
Learn More

Ethylene glycol diacetate, 99%

Product Code. 10347930 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
10 L
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10347930 2.5 L Glass bottle 2.5 L
10022400 500 mL Glass bottle 500mL
10127353 10 L Plastic drum 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10347930 Supplier Thermo Scientific Acros Supplier No. 176220025
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 111-55-7 | C6H10O4 | 146.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-55-7
Molecular Formula C6H10O4
Molecular Weight (g/mol) 146.14
MDL Number MFCD00008718
InChI Key JTXMVXSTHSMVQF-UHFFFAOYSA-N
Synonym ethylene glycol diacetate, 1,2-diacetoxyethane, glycol diacetate, ethylene diacetate, ethanediol diacetate, 1,2-ethanediol, diacetate, 1,2-ethanediol diacetate, ethylene glycol, diacetate, ethylene diethanoate, aptex donor h-plus
PubChem CID 8121
IUPAC Name 2-acetyloxyethyl acetate
SMILES CC(=O)OCCOC(=O)C

Specifications

Melting Point -41°C
Density 1.1200g/mL
Boiling Point 186°C to 187°C
Flash Point 82°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CO2CH2CH2O2CCH3
Refractive Index 1.4142 to 1.4162
Quantity 2.5 L
Beilstein 02, 142
Merck Index 15, 3853
Specific Gravity 1.12
Solubility Information Solubility in water: 160g/L (20°C). Other solubilities: soluble in alcohol,ether and benzene,miscible with ethanol and acetone
Viscosity 2.58 mm2/s (20°C)
Formula Weight 146.14
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Ethylene glycol diacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.