missing translation for 'onlineSavingsMsg'
Learn More

Ethylene glycol-monophenyl ether, Dr. Ehrenstorfer

Product Code. 30316206
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30316206 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30316206 Supplier Dr. Ehrenstorfer Supplier No. DREC13328300100MG
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenoxyethanol
CAS 122-99-6
Analyte or Component Names Phenoxyethanol
Shipping Condition +20°C
Molecular Formula C8 H10 O2
Certifications/Compliance ISO 17034
Synonym Phenoxyethanol, (2-Hydroxyethoxy)benzene, 1-Hydroxy-2-phenoxyethane, 2-Hydroxyethyl phenyl ether, 2-Phenoxyethanol, 2-Phenoxyethyl alcohol, Arosol, Dalpad A, Dowanol EP, Dowanol EPh, Emeressence 1160, Ethylene glycol monophenyl ether, Ethylene glycol phenyl ether, H 4644, Hisolve EPH, NSC 1864, Newpol EFP, PHE, PHE (alcohol), PHE-G, PHE-S, Phenova, Phenoxethol, Phenoxetol, Phenoxyethyl alcohol, Phenyl cellosolve, Plastilit DS 3431, Rokafenol F 1, Sepicide LD, beta-Hydroxyethyl phenyl ether, beta-Phenoxyethanol, beta-Phenoxyethyl alcohol
InChI Formula InChI=1S/C8H10O2/c9-6-7-10-8-4-2-1-3-5-8/h1-5,9H,6-7H2
SMILES OCCOc1ccccc1
IUPAC Name 2-phenoxyethanol
Molecular Weight (g/mol) 138.16
Quantity 100 mg
Formula Weight 138.0681 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.