missing translation for 'onlineSavingsMsg'
Learn More

Ethylene oxide 50000 μg/mL in Dichloromethane, Dr. Ehrenstorfer

Product Code. 30327202
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30327202 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30327202 Supplier Dr. Ehrenstorfer Supplier No. DREA15786500DI50K
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Oxirane
CAS 75-21-8
Analyte or Component Names Oxirane
Shipping Condition -18°C
Molecular Formula C2 H4 O
Concentration 50000 μg/mL
Certifications/Compliance ISO 17034
Synonym Oxirane, Ethylene oxide
InChI Formula InChI=1S/C2H4O/c1-2-3-1/h1-2H2
SMILES C1CO1
IUPAC Name oxirane
Solvent or Matrix Dichloromethane
Molecular Weight (g/mol) 44.05
Quantity 1 mL
Formula Weight 44.0262 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.