missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Terephthalate Cyclic Pentamer, TRC

Product Code. 30009415
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009415 5 mg 5mg
30004173 2.5 mg 2.5mg
30005960 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009415 Supplier Toronto Research Chemicals Supplier No. TRCE9181855MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16104-97-5
Molecular Formula C50 H40 O20
Molecular Weight (g/mol) 960.841
Synonym 3,6,13,16,23,26,33,36,43,46-Decaoxahexacyclo[46.2.2.28,11.218,21.228,31.238,41]hexaconta-8,10,18,20,28,30,38,40,48,50,51,53,55,57,59-pentadecaene-2,7,12,17,22,27,32,37,42,47-decone, Terephthalic Acid, Pentamol. Cyclic Ethylene Ester, Ethylene Glycol, Pentamol. Cyclic Terephthalate
SMILES O=C1OCCOC(=O)c2ccc(cc2)C(=O)OCCOC(=O)c3ccc(cc3)C(=O)OCCOC(=O)c4ccc(cc4)C(=O)OCCOC(=O)c5ccc(cc5)C(=O)OCCOC(=O)c6ccc1cc6

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C50H40O20/c51-41-31-1-2-32(4-3-31)42(52)62-23-24-64-44(54)34-9-11-36(12-10-34)46(56)66-27-28-68-48(58)38-17-19-40(20-18-38)50(60)70-30-29-69-49(59)39-15-13-37(14-16-39)47(57)67-26-25-65-45(55)35-7-5-33(6-8-35)43(53)63-22-21-61-41/h1-20H,21-30H2
Formula Weight 960.211
Percent Purity >95%
Chemical Name or Material Ethylene Terephthalate Cyclic Pentamer
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.