missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylicin 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Ethylicin |
| CAS | 682-91-7 |
| Analyte or Component Names | Ethylicin |
| Shipping Condition | +20°C |
| Molecular Formula | C4 H10 O2 S2 |
| Concentration | 100 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | Ethanesulfonothioic acid, S-ethyl ester, Ethanesulfonic acid, thio-, S-ethyl ester (6CI,7CI,8CI), Ethanethiol, ethanesulfonate (6CI,8CI), Ethanethiosulfonic acid ethyl ester, Ethyl ethanethiosulfonate, NSC 22422, Pseudoallicin 150, S-Ethyl ethanethiosulfonate, Ethylicin |
| InChI Formula | InChI=1S/C4H10O2S2/c1-3-7-8(5,6)4-2/h3-4H2,1-2H3 |
| SMILES | CCSS(=O)(=O)CC |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?