missing translation for 'onlineSavingsMsg'
Learn More

Ethylmagnesium bromide, 0.9M in tert-Butyl methyl ether, AcroSeal™, ACROS Organics™

Product Code. 10736633 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
800 mL
Unit Size:
800mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10736633 800 mL 800mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10736633 Supplier Thermo Scientific Acros Supplier No. 347288000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Specifications

Color Brown
CAS 925-90-6, 1634-04-4
Molecular Formula C2H5BrMg
Linear Formula C2H5MgBr
MDL Number MFCD00000043
Concentration 13 to 16%
Fieser 02,205; 02,205
Density 0.8400g/mL
Synonym ethylmagnesium bromide, ethyl magnesium bromide, magnesium, bromoethyl, ethylmagnesiumbromide, bromoethylmagnesium, ethylmagnesium bromide solution, 1.0 m in thf, ethylmagnesium bromide solution, 3.0 m in diethyl ether, etmgbr, ethylmagnesiumbromid, ethylmagnesium bromid
InChI Key TWTWFMUQSOFTRN-UHFFFAOYSA-M
SMILES CC[Mg]Br
IUPAC Name bromo(ethyl)magnesium
Molecular Weight (g/mol) 133.27
PubChem CID 101914
Formula Weight 133.27
Packaging AcroSeal™ Glass Bottle
Quantity 800 mL
Physical Form Liquid
Chemical Name or Material Ethylmagnesium bromide, 0.9M Solution in tert-Butyl Methyl Ether
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.