missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Etoxazole-amino hydrochloride, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Etoxazole Amino Hydrochloride |
| Analyte or Component Names | Etoxazole Amino Hydrochloride |
| Shipping Condition | +20°C |
| Molecular Formula | C21 H25 F2 N O3 . H Cl |
| Certifications/Compliance | ISO/IEC 17025 |
| Synonym | Etoxazole-amino hydrochloride, 2 amino-2-(4-tert-butyl-2-ethoxyphenyl)ethyl 2',6'-difluorobenzoate hydrochloride, Etoxazole Amino Hydrochloride |
| InChI Formula | InChI=1S/C21H25F2NO3.ClH/c1-5-26-18-11-13(21(2,3)4)9-10-14(18)17(24)12-27-20(25)19-15(22)7-6-8-16(19)23;/h6-11,17H,5,12,24H2,1-4H3;1H |
| SMILES | Cl.CCOc1cc(ccc1C(N)COC(=O)c2c(F)cccc2F)C(C)(C)C |
| IUPAC Name | [2-amino-2-(4-tert-butyl-2-ethoxy-phenyl)ethyl] 2,6-difluorobenzoate;hydrochloride |
| Molecular Weight (g/mol) | 413.88585 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?