missing translation for 'onlineSavingsMsg'
Learn More

Etravirine, MedChemExpress

Product Code. 16478248
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16478248 10 mg 10mg
16454137 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16478248 Supplier Medchem Express Supplier No. HY90005
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Etravirine is a non-nucleoside reverse transcriptase inhibitor (NNRTI) used for the treatment of HIV.

Etravirine is a non-nucleoside reverse transcriptase inhibitor (NNRTI) used for the treatment of HIV.

Specifications

Chemical Name or Material Etravirine
Physical Form Powder
CAS 269055-15-4
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C20H15BrN6O
For Use With (Application) COVID-19-anti-virus
Synonym R165335 TMC125
Solubility Information DMSO : 50 mg/mL (114.87 mM; Need ultrasonic)
SMILES N#CC1=CC(C)=C(C(C)=C1)OC2=NC(NC3=CC=C(C=C3)C#N)=NC(N)=C2Br
Molecular Weight (g/mol) 435.28
Formula Weight 435.28
Percent Purity 98.8%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.