missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ETS1 associated protein II Antibody (TDP2/1258), Novus Biologicals™
Mouse Monoclonal Antibody
265.00 € - 474.00 €
Specifications
| Antigen | ETS1 associated protein II |
|---|---|
| Clone | TDP2/1258 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 1-2 μg/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL, Immunofluorescence 0.5-1ug/mL |
| Applications | Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18691528
|
Novus Biologicals
NBP2-50070SS |
0.02 mg |
265.00 €
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18649898
|
Novus Biologicals
NBP2-50070-0.1mg |
0.1 mg |
474.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ETS1 associated protein II Monoclonal antibody specifically detects ETS1 associated protein II in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), ImmunofluorescenceSpecifications
| ETS1 associated protein II | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Signal Transduction, Transcription Factors and Regulators | |
| 10 mM PBS with 0.05% BSA | |
| AD022, dJ30M3.3, EAP2, EAPII, EC 3.1.4.-, ETS1-associated protein 2, ETS1-associated protein II, MGC111021,5'-Tyr-DNA phosphodiesterase, MGC9099,5'-tyrosyl-DNA phosphodiesterase, TRAF and TNF receptor associated protein, TTRAPTRAF and TNF receptor-associated protein, Tyr-DNA phosphodiesterase 2, tyrosyl-DNA phosphodiesterase 2 | |
| Recombinant full-length human ETS1 associated protein II protein (Uniprot: O95551) | |
| Primary | |
| Store at 4°C. |
| TDP2/1258 | |
| Flow Cytometry 1-2 μg/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL, Immunofluorescence 0.5-1ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| O95551 | |
| 51567 | |
| IgG2b κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title