missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ETS2 Mouse anti-Human, Clone: OTI1H4, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505372S
This item is not returnable.
View return policy
Description
ETS transcriptions factors, such as ETS2, regulate numerous genes and are involved in stem cell development, cell senescence and death, and tumorigenesis. The conserved ETS domain within these proteins is a winged helix-turn-helix DNA-binding domain that recognizes the core consensus DNA sequence GGAA/T of target genes.Specifications
| ETS2 | |
| Monoclonal | |
| Unconjugated | |
| ETS2 | |
| ETS2IT1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 2114 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1H4 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P15036 | |
| ETS2 | |
| Human recombit protein fragment corresponding to amino acids 90-349 of human ETS2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction