missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ETV1 Recombinant Rabbit Monoclonal Antibody (8O4N2)

Rabbit Recombinant Monoclonal Antibody
176.00 € - 436.00 €
Specifications
| Antigen | ETV1 |
|---|---|
| Clone | 8O4N2 |
| Concentration | 1 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ChIP Assay, ELISA, Immunoprecipitation, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30283332
|
Invitrogen™
MA557939 |
20 μL |
176.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30283991
![]() |
Invitrogen™
MA557940 |
100 μL |
436.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Immunogen sequence: MDGFYDQQVP YMVTNSQRGR NCNEKPTNVR KRKFINRDLA HDSEELFQDL SQLQETWLAE AQVPDNDEQF VPDYQAESLA FHGLPLKIKK EPHSPCSEIS.
ETV1: ets variant gene 1. Several members of the Ets gene family encode sequence-specific DNA binding proteins that recognize DNA sequences with a centrally located 5'-GGAA-3' element. All of the Ets proteins recognize the same central core sequence but each protein interacts with unique sequences that flank this core. ER81 (also designated ETV1) binds the motif 5'-CGGAA/T-3'. ER81 is highly expressed in brain, testis, lung and heart. ER81 is also moderately expressed in spleen, pancreas, colon and small intestine. During development, ER81 displays a unique expression patterns which suggests that this transcriptional factor might play an important role in organogenesis. ERK-1 activates ER81 transcriptional activity while MAPKAP kinase 2 inhibits ER81.Specifications
| ETV1 | |
| 1 mg/mL | |
| ChIP Assay, ELISA, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P41164, P50549 | |
| 14009, 2115, 362733 | |
| Synthetic peptide. | |
| Antibody |
| 8O4N2 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| Etv1 | |
| DKFZp781L0674; Er81; ets related protein 81; ETS transcription factor Er81; ETS translocation variant 1; ets variant 1; ets variant gene 1; ETS variant transcription factor 1; ets-related protein 81; Etsrp81; Etv1; MGC104699; MGC120533; MGC120534; MGC137631 protein | |
| Etv1 | |
| Primary | |
| Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title