missing translation for 'onlineSavingsMsg'
Learn More

EU Drinking Water Directive PFAS Mixture 10 μg/mL in Methanol:Water, Dr. Ehrenstorfer

Product Code. 30316181
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30316181 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30316181 Supplier Dr. Ehrenstorfer Supplier No. DREA30000051MW
Big Savings for Better Separation
Use promo code 27992
View all promotions
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Perfluorobutanesulfonic acid∣Perfluorobutanoic acid∣Perfluorodecane sulfonic acid∣Perfluorodecanoic acid∣Perfluorododecanesulfonic Acid∣Perfluorododecanoic acid∣Perfluoroheptanesulfonic acid∣Perfluoroheptanoic acid∣Perfluorohexanesulfonic acid∣Perfluorohexanoic acid∣Perfluorononanesulfonic Acid∣Perfluorononanoic acid∣Perfluorooctanesulfonic acid∣Perfluorooctanoic acid∣Perfluoropentanesulfonic Acid∣Perfluoropentanoic acid∣Perfluorotridecanesulfonic Acid∣Perfluorotridecanoic acid∣Perfluoroundecanesulfonic Acid∣Perfluoroundecanoic acid
CAS 375-73-5, 375-22-4, 335-77-3, 335-76-2, 79780-39-5, 307-55-1
Analyte or Component Names Perfluorobutanesulfonic acid∣Perfluorobutanoic acid∣Perfluorodecane sulfonic acid∣Perfluorodecanoic acid∣Perfluorododecanesulfonic Acid∣Perfluorododecanoic acid∣Perfluoroheptanesulfonic acid∣Perfluoroheptanoic acid∣Perfluorohexanesulfonic acid∣Perfluorohexanoic acid∣Perfluorononanesulfonic Acid∣Perfluorononanoic acid∣Perfluorooctanesulfonic acid∣Perfluorooctanoic acid∣Perfluoropentanesulfonic Acid∣Perfluoropentanoic acid∣Perfluorotridecanesulfonic Acid∣Perfluorotridecanoic acid∣Perfluoroundecanesulfonic Acid∣Perfluoroundecanoic acid
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Methanol/Water 96/4
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Perfluorobutanesulfonic acid 10 μg/mL∣Perfluorobutanoic acid 10 μg/mL∣Perfluorodecane sulfonic acid 10 μg/mL∣Perfluorodecanoic acid 10 μg/mL∣Perfluorododecanesulfonic Acid 10 μg/mL∣Perfluorododecanoic acid 10 μg/mL∣Perfluoroheptanesulfonic acid 10 μg/mL∣Perfluoroheptanoic acid 10 μg/mL∣Perfluorohexanesulfonic acid 10 μg/mL∣Perfluorohexanoic acid 10 μg/mL∣Perfluorononanesulfonic Acid 10 μg/mL∣Perfluorononanoic acid 10 μg/mL∣Perfluorooctanesulfonic acid 10 μg/mL∣Perfluorooctanoic acid 10 μg/mL∣Perfluoropentanesulfonic Acid 10 μg/mL∣Perfluoropentanoic acid 10 μg/mL∣Perfluorotridecanesulfonic Acid 10 μg/mL∣Perfluorotridecanoic acid 10 μg/mL∣Perfluoroundecanesulfonic Acid 10 μg/mL∣Perfluoroundecanoic acid 10 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.