missing translation for 'onlineSavingsMsg'
Learn More

Standard Color Solution Y, Yellow, EP Grade, Reagecon™
BIOPHARMA

Product Code. 11894232 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
125 mL
Unit Size:
125mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11894232 125 mL 125mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11894232 Supplier Reagecon Supplier No. EP706
  • ONLINE EXCLUSIVE PRICE
    254.95 €
    147.93 € / 125mL
    Save 107.02 €
    41% Off
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Used to calibrate, control, qualify and validate color measurement instruments. Standard Color Solution Y, Yellow, EP Grade, Reagecon™ is a ready-to-use colorimetric solution produced in accordance with Eu. Ph. Chap 2 from a precise combination of Primary Color Solutions.

Ready-to-use colorimetric standard presented in a high-quality, tamper-evident bottle. EP Standard Color Solutions are prepared from the EP Primary Color Solutions (yellow, red and blue) and can be used to prepare EP Reference Color Solutions.
  • Manufactured according to European Pharmacopoeia Chapter 2 guidelines
  • Balance Certificate of Calibration issued in accordance with ISO 17025
  • Prepared gravimetrically on a weight/weight basis using OIML traceable weights
  • Concentration verified using a high-performance spectrophotometer calibrated using ISO Guide 34 standards
  • Color Standard Y = 2.4 Yellow + 0.6 Red + 7.0 Hydrochloric Acid (10g HCl/L)

Specifications

Description European Pharmacopoeia Standard Color Solution Y
Packaging Tamper evident
Certificate of Analysis Yes
CAS 7705-08-0, 7646-79-9, 7647-01-0
Analyte or Component Names Iron chloride, Cobalt chloride, Hydrochloric acid
Physical Form Liquid
Color Yellow
Element Iron Chloride, Cobalt Chloride, Hydrochloric Acid
Quantity 125 mL
Grade Eu. Ph. Chapter 2
compliance-icons compliance-icons compliance-icons
Product Identifier
  • STANDARD SOLUTION Y (YELLOW)
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
  • Reproductive toxicity Category 1B
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H350i-May cause cancer by inhalation.
  • H360F-May damage fertility.
  • H411-Toxic to aquatic life with long lasting effects.
  • H319-Causes serious eye irritation.
  • EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contain: hydrochloric acid 37%, iron trichloride,cobalt dichloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.