missing translation for 'onlineSavingsMsg'
Learn More

Europium for AAS 1g/L HNO3 matrix, Fisher chemical™
BIOPHARMA

Product Code. 12903155 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Bottle
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
12903155 100 mL Bottle 100mL
12913155 500 mL Bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12903155 Supplier Fisher Chemical Supplier No. B 018 100
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Chemical Identifiers

CAS 7440-53-1
Molecular Formula Eu
Molecular Weight (g/mol) 151.96
MDL Number MFCD00010992
InChI Key OGPBJKLSAFTDLK-UHFFFAOYSA-N
PubChem CID 23981
ChEBI CHEBI:32999
IUPAC Name europium
SMILES [Eu]

Specifications

Packaging Bottle
Physical Form Liquid
Concentration 1 g/L ±0.002 g/L
Type Flame Single-element Standard Solution
Solvent or Matrix 2% HNO3
Quantity 100 mL
Chemical Name or Material Europium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.