missing translation for 'onlineSavingsMsg'
Learn More

Europium Standard for ICP/MS - Claritas PPT Grade, SPEX CertiPrep™

Product Code. 15235926 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
125 mL
Unit Size:
125mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15235926 125 mL 125mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15235926 Supplier SPEX Certiprep Supplier No. CLEU21BY

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 7440-53-1
Molecular Formula Eu
MDL Number MFCD00010992
InChI Key OGPBJKLSAFTDLK-UHFFFAOYSA-N
SMILES [Eu]
IUPAC Name europium
Solvent or Matrix 2% HNO3
Molecular Weight (g/mol) 151.96
PubChem CID 23981
Quantity 125 mL
ChEBI CHEBI:32999
Concentration or Composition (by Analyte or Components) 1mg/L
Chemical Name or Material Europium Standard for ICP/MS - Claritas PPT Grade
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.