Learn More
R&D Systems™ Europium Streptavidin MIDI
Product Code. 18081104 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
400µL
Product Code. Quantity Unit Size
18081104 400 μL 400µL
1 options
Product Code. 18081104 Supplier R&D Systems™ Supplier No. 12200120
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
A Novus product, part of R&D Systems. Europium (Eu) Streptavidin conjugates are manufactured using covalent attachment of Streptavidin to the specially treated surface of 200nm Eu particles.
  • Highly stable, easy to use conjugates
  • Resistant to aggregation
  • No sonication, shaking or vortexing required
  • No need for blocking solution
  • Possibility to work with low conjugate concentrations
  • Conjugates can be run on an immunochromatographic assay as well as a microwell-based assay
Kit Contents:
  • 1 Midi vial of 200 nmEuropium - Streptavidin conjugate
TRUSTED_SUSTAINABILITY
Specifications
Quantity 400 μL
Content And Storage Storage of components varies. See protocol for specific instructions.
Product Line Novus