missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Eutech™ Ion 6+ meter
Quick and easy measurement with this ion-selective meter.
496.00 € - 593.00 €
Specifications
Accuracy (ISE) | ± 1% full scale |
---|---|
Accuracy (mV) | 1 mV & ±2 mV (beyond ±200 mV) |
Accuracy (pH) | ± 0.01 |
Accuracy (Temperature) | ±0.5°C |
Battery Type | 4 AAA x 1.4V |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
11549174
|
Thermo Scientific™
01X256409 |
Eutech Ion 6+ Ion/pH/mV Meter with ATC Electrode ECPHTEM01P, Rubber Armour/Stand |
496.00 €
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11559174
|
Thermo Scientific™
01X256410 |
Eutech Ion 6+ Ion/pH/mV Meter with Single Junction pH Electrode ECFC7252101B, ATC Electrode ECPHTEM01P, & pH Carrying Kit (Includes 1 x pH 4.01, pH 7.00 Buffer Solutions, Storage Solution & 1 Deionised (Rinse) water (60 ml Each), Rubber Armour/Stand |
593.00 €
Each |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Descripción
Quick and easy measurement with this ion-selective meter. Applications for the Eutech™ Ion 6+ meter include: ISE (Use with any ISE with BNC connection), water quality testing, environmental/agricultural testing, laboratories and schools, and in industrial settings.
Features
- Direct Ion Concentration for use with most mono- or di-valent Ion Selective Electrodes (ISE)
- mV measurement mode allows you to check performance and calibration of your ISE
- Multi-Point Calibration (up to 5 points) with Auto-Buffer Recognition - choice of USA, NIST or pure water buffer standards with ±0.01 pH accuracy
- Automatic Temperature Compensation (ATC) for the highest accuracy under fluctuating temperatures
- Non-Volatile Memory ensures data are retained even meter powers off
- Hold Function momentarily freezes reading for easy viewing and recording
- Auto-Power Off after 17 minutes saves battery power after non-use
- Self-Diagnostic with message codes for easy troubleshooting
- Large Custom LCD provides optimum viewing even at a distance
- Protective Rubber Boot shields meter from accidental knocks and features a sturdy built-in stand for easy bench-top operation
- Rugged Carrying Case (Optional) provides handy on-site meter calibration and measurement
- pH Buffer Options: 1.68, 4.01, 7.00, 10.01, 12.45 (USA), 1.68, 4.01, 6.86, 9.18, 12.45 (NIST), 4.10, 6.97 (Pb)
Especificaciones
± 1% full scale | |
± 0.01 | |
4 AAA x 1.4V | |
Up to 5 | |
Ion Concentration: 0.01 to 0.99 ppm, 1.0 to 199.9 ppm, 200 to 1999 ppm | |
0.00 to 14.00 | |
0.01 | |
0.1 mV and ±0.2 mV (within ±199.9 mV) |
1 mV & ±2 mV (beyond ±200 mV) | |
±0.5°C | |
Up to 5 points | |
Ion-Selective Meter | |
± 500 mV | |
0.01 ppm, 0.1 ppm, 1 ppm | |
0°C to 100.0°C | |
0.1°C |