missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eutech pH Buffer Sachets
Description
pH Buffer Sachets are NIST-traceable sachets for quick, accurate and convenient calibrations. Simply insert electrode into sachet pack, calibrate, rinse and discard sachet – no separate container needed.
Features:
- pH Buffer Sachets -20 ml
- NIST Traceable
Specifications
Specifications
| Chemical Name or Material | pH Buffer Sachet |
| For Use With (Equipment) | pH Meter and Electrode |
| pH | 7 |
| Quantity | 20 x 20 mL |
| Solution Type | Buffer |
| Certifications/Compliance | NIST Traceable |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?