missing translation for 'onlineSavingsMsg'
Learn More

Eutech™ pH Buffer Tablets

Product Code. 12631125
Click to view available options
pH:
4
7
10
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 12631125

Brand: Thermo Scientific Eutech 01X038201

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Eutech™ offers pH buffer tablets for use with pH meters and electrodes

Eutech™ pH buffer tablets provide a quick, convenient way to prepare pH buffers from tablets for pH calibrations. Each box contains 100 pH buffer tablets.

Features:

  • 10 strips x 10 tablets per box
TRUSTED_SUSTAINABILITY

Specifications

Color Red
For Use With (Application) pH calibration
For Use With (Equipment) pH Meter and Electrode
pH 4
Physical Form Tablet
Quantity 100 Tablets
Solution Type Buffer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.