Description
Retsch PP 25 Pellet Press is a compact benchtop unit with particularly simple and safe operation.
- Produces high quality, stable pellets
- Easy and safe operation
- Several pressing tools available
- Can be evacuated
- Compact benchtop unit
Specifications
Specifications
| Product Type | Evacuable Pressing Tools |
| Diameter (Metric) | 32 mm dia. Pellets |
| For Use With (Equipment) | Pellet Press PP 25 |
| Weight (Metric) | 1 kg |