Learn More
Tocris Bioscience™ Everolimus
GREENER_CHOICE
mTOR inhibitor;immunosuppressant
Brand:  Tocris Bioscience™ 6188/10
Encompass
Product Code. 15756825
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Everolimus is a mTOR inhibitor. Inhibits tumor proliferation in vitro and in vivo. Inhibits VEGF-induced HUVEC proliferation in vitro, but does not affect tumor vascular permeability in mouse models. Immunosuppressant. Activates autophagy.
Specifications
Specifications
mTOR Inhibitor, Immunosuppressant
159351-69-6
958.22g/mol
mTOR Inhibitors
mTOR
C53H83NO14
Store at -20°C