missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EVL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13484-1-AP-20UL
This item is not returnable.
View return policy
Description
Ena/VASP proteins are actin-associated proteins involved in a range of processes dependent on cytoskeleton remodeling and cell polarity such as axon guidance and lamellipodial and filopodial dynamics in migrating cells. EVL enhances actin nucleation and polymerization.Specifications
| EVL | |
| Polyclonal | |
| Unconjugated | |
| EVL | |
| Ena/VASP like protein, Enah/Vasp like, EVL, RNB6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14026, 51466, 79115 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.63 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O08719, P70429, Q9UI08 | |
| EVL | |
| EVL Fusion Protein Ag4302 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction