Learn More
Thermo Scientific™ Evodiamine
70.30 € - 124.00 €
Specifications
| Description | Off-white to pale yellow |
|---|---|
| Content And Storage | -30°C to -10°C |
| Format | Solid |
| For Use With (Application) | A vanilloid receptor agonist that induces apoptosis in leukemia U937 cells |
| CAS | 518-17-2 |
| Molecular Formula | C19H17N3O |
| Formula Weight | 303.40 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15465069
|
Thermo Scientific™
J62771.MD |
250 mg |
124.00 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15455069
|
Thermo Scientific™
J62771.MC |
100 mg |
70.30 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Off-white to pale yellow | |
| Solid | |
| 518-17-2 | |
| 303.40 |
| -30°C to -10°C | |
| A vanilloid receptor agonist that induces apoptosis in leukemia U937 cells | |
| C19H17N3O |
Safety and Handling
Recommended Storage : Store at -20°C
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.