Learn More
Isofield™ Exa 230 Validated Sterile Earloop Cleanroom Facemask with Clip - Grade A/B Compatible
BIOPHARMA
Product Code. 30178865
Change view
Click to view available options
Size:
Wide
Unit Size:
Pack of 600
Product Code. Size Unit Size
30178865 Wide Pack of 600
1 options
Product Code. 30178865 Supplier Isofield™ Supplier No. 60011
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
Estimated Shipment: 18-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Extra wide validated sterile facemask. The Exa is also fitted with a clip to adapt to your SOP, and is made with gamma compatible materials to ensure low/no odor in use. The Exa also has excellent bacterial and particle filtration efficiency and is GMP Grade A compatible.
  • Validated sterile earloop cleanroom facemask with clip
  • 230 mm length with malleable nose strip
  • 3-layer non-woven configuration
  • Highly efficient bacterial and particle filtration
  • Gamma irradiation, minimum 25 kGy
  • Sterility Assurance Level 10-6
  • Particle Shedding (Helmke Drum) Category I Individually packed pouches printed with IPA resistant ink to maintain traceability
Specifications
Product Type Cleanroom Face Mask
Color White
Fastening Type Earloops
Material Polyethylene, Polypropylene
Size Wide
Sterility Sterile
Style Pleated