missing translation for 'onlineSavingsMsg'
Learn More

Exatecan mesylate, MedChemExpress

Product Code. 16446256
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16446256

Brand: Medchem Express HY13631A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Exatecan mesylate (DX8951f) is a DNA topoisomerase I inhibitor, with an IC50 of 2.2 μM (0.975 μg/mL). Exatecan mesylate can be used in cancer research.

Exatecan mesylate (DX8951f) is a DNA topoisomerase I inhibitor, with an IC50 of 2.2 μM (0.975 μg/mL). Exatecan mesylate can be used in cancer research.

Specifications

Chemical Name or Material Exatecan mesylate
CAS 169869-90-3
Color Light Green
Purity Grade Notes Research
Molecular Formula C25H26FN3O7S
Quantity 100 mg
Synonym DX8951f
Solubility Information DMSO : 11.76 mg/mL (22.12 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 80°C) (insoluble)
SMILES O=S(C)(O)=O.O=C1[C@](O)(CC)C2=C(CO1)C(N3CC4=C5C6=C(CC[C@@H]5N)C(C)=C(F)C=C6N=C4C3=C2)=O
Molecular Weight (g/mol) 531.55
Formula Weight 531.55
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.