missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EXOC2 Antibody (0H2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43031
This item is not returnable.
View return policy
Description
EXOC2 Monoclonal antibody specifically detects EXOC2 in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| EXOC2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| 0H2 | |
| Western Blot 1:2500 - 1:10000, ELISA, Immunohistochemistry 1:200 - 1:800, Immunocytochemistry/ Immunofluorescence 1:10 - 1:100, Immunoprecipitation 1:500 - 1:1000 | |
| exocyst complex component 2, Exocyst complex component Sec5, FLJ11026, SEC5, SEC5L1, SEC5-like 1, SEC5-like 1 (S. cerevisiae), Sec5p | |
| exocyst complex component 2 - (Uniprot# Q96KP1) | |
| 100 μg | |
| Signal Transduction | |
| 55770 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur